Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGIS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000961.3 | NP_000952.1 | 3, 4 | 425 | ||
GenBank mRNA | AF297048.1 | - | 3, 4 | 371 | ||
AF297049.1 | - | 3, 4 | 371 | |||
AF297050.1 | - | 3, 4 | 371 | |||
AF297051.1 | - | 3, 4 | 371 | |||
AF297052.1 | - | 3, 4 | 371 | |||
BC101809.1 | - | 3, 4 | 446 | 8069315 | ||
BC101811.1 | - | 3, 4 | 446 | 8069317 | ||
D38145.1 | - | 3, 4 | 398 |