Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000962.3 | NP_000953.2 | 11 | 2847 | ||
NM_001271164.1 | NP_001258093.1 | 10 | 2703 | |||
NM_001271165.1 | NP_001258094.1 | 10 | 2796 | |||
NM_001271166.1 | NP_001258095.1 | 11 | 2962 | |||
NM_001271367.1 | NP_001258296.1 | 11 | 2707 | |||
NM_001271368.1 | NP_001258297.1 | 12 | 3184 | |||
NM_080591.2 | NP_542158.1 | 11 | 2736 | |||
XM_005252105.3 | XP_005252162.1 | 12 | 3022 | |||
XM_011518875.2 | XP_011517177.1 | 12 | 3283 | |||
XM_011518876.2 | XP_011517178.1 | 12 | 6893 | |||
XM_017014929.1 | XP_016870418.1 | 14 | 3257 | |||
GenBank mRNA | U63846.1 | - | 1 | 1023 |