Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033.4 | NP_001024.1 | 14 | 1855 | ||
NM_001318064.1 | NP_001304993.1 | 13 | 1677 | |||
NM_001318065.1 | NP_001304994.1 | 8 | 1202 | |||
XM_011520278.1 | XP_011518580.1 | 8 | 1155 | |||
GenBank mRNA | AK222786.1 | - | 14 | 1735 | ||
AK297988.1 | - | 14 | 1600 | |||
AK301891.1 | - | 8 | 995 | |||
AK301922.1 | - | 8 | 1037 | |||
AK303394.1 | - | 13 | 1521 | |||
BC006498.2 | - | 14 | 1686 | 2989344 | ||
X59543.1 | - | 14 | 1714 | |||
X59617.1 | - | 14 | 1708 |