Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL9A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001851.4 | NP_001842.3 | 15, 16 | 1365 | ||
NM_078485.3 | NP_511040.2 | 9, 10 | 633 | |||
XM_011535429.2 | XP_011533731.1 | 15, 16 | 1365 | |||
XM_011535430.2 | XP_011533732.1 | 9, 10 | 631 | |||
XM_017010246.1 | XP_016865735.1 | 12, 13 | 722 | |||
XM_017010247.1 | XP_016865736.1 | 6, 7 | 578 | |||
GenBank mRNA | BC008620.1 | - | 9, 10 | 560 | ||
BC063646.1 | - | 9, 10 | 607 | 4914907 | ||
X54412.1 | - | 15, 16 | 1337 |