Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001091.3 | NP_001082.2 | 2 | 847 | ||
NM_001272072.1 | NP_001259001.1 | 2 | 847 | |||
XM_017011944.1 | XP_016867433.1 | 4 | 1055 | |||
XM_017011945.1 | XP_016867434.1 | 4 | 1085 | |||
XM_017011946.1 | XP_016867435.1 | 2 | 1011 | |||
XM_017011947.1 | XP_016867436.1 | 2 | 825 | |||
GenBank mRNA | AK092514.1 | - | 3 | 662 | ||
AK296638.1 | - | 2 | 807 | |||
AK307278.1 | - | 3 | 532 | |||
AK314806.1 | - | 2 | 805 | |||
BC014093.2 | - | 2 | 800 | 4547491 | ||
BX648159.1 | - | 4 | 4970 | |||
U11862.1 | - | 2 | 847 | |||
U11863.1 | - | 2 | 822 |