Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALOX15B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039130.1 | NP_001034219.1 | 9 | 1445 | ||
NM_001039131.1 | NP_001034220.1 | 9 | 1445 | |||
NM_001141.2 | NP_001132.2 | 10 | 1532 | |||
GenBank mRNA | AF468051.1 | - | 10 | 1416 | ||
AF468052.1 | - | 9 | 1329 | |||
AF468053.1 | - | 9 | 1329 | |||
AF468054.1 | - | 10 | 1416 | |||
AK292779.1 | - | 10 | 1528 | |||
AK298095.1 | - | 10 | 1538 | |||
AK300554.1 | - | 10 | 1516 | |||
BC035217.1 | - | 10 | 1474 | 4778890 | ||
BC063647.1 | - | 10 | 1488 | 4804186 | ||
U78294.1 | - | 10 | 1487 |