Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANK2 SIRNA Assays ›
Gene Symbol:
ANK2
Gene Name:
ankyrin 2, neuronal
Gene Aliases:
ANK-2, LQT4, brank-2
Chromosome Location:
Chr.4: 112706083 - 113383740 on Build GRCh38
UniGene ID:
Hs.620557
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
106390
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127493.1
NP_001120965.1
27
2991
NM_001148.4
NP_001139.3
26
2933
NM_020977.3
NP_066187.2
26
2933
XM_005262945.2
XP_005263002.1
26
3031
XM_017008067.1
XP_016863556.1
27
3854
XM_017008068.1
XP_016863557.1
28
4095
XM_017008069.1
XP_016863558.1
26
3829
XM_017008070.1
XP_016863559.1
27
4040
XM_017008071.1
XP_016863560.1
28
3023
XM_017008072.1
XP_016863561.1
27
2999
XM_017008073.1
XP_016863562.1
26
3031
XM_017008074.1
XP_016863563.1
27
2969
XM_017008075.1
XP_016863564.1
27
2969
XM_017008076.1
XP_016863565.1
26
3031
XM_017008077.1
XP_016863566.1
26
2945
XM_017008078.1
XP_016863567.1
26
3031
XM_017008079.1
XP_016863568.1
25
2860
XM_017008080.1
XP_016863569.1
26
2945
XM_017008081.1
XP_016863570.1
26
3031
XM_017008082.1
XP_016863571.1
27
3172
XM_017008083.1
XP_016863572.1
27
3172
XM_017008084.1
XP_016863573.1
26
3031
XM_017008085.1
XP_016863574.1
27
3028
XM_017008086.1
XP_016863575.1
27
3028
XM_017008087.1
XP_016863576.1
27
3172
XM_017008088.1
XP_016863577.1
28
2966
XM_017008089.1
XP_016863578.1
26
3175
XM_017008090.1
XP_016863579.1
27
3172
XM_017008091.1
XP_016863580.1
25
2932
XM_017008092.1
XP_016863581.1
27
2969
XM_017008093.1
XP_016863582.1
28
2966
XM_017008094.1
XP_016863583.1
27
3028
XM_017008095.1
XP_016863584.1
26
2945
XM_017008096.1
XP_016863585.1
28
2966
XM_017008097.1
XP_016863586.1
26
2870
XM_017008098.1
XP_016863587.1
27
2969
XM_017008099.1
XP_016863588.1
28
2966
XM_017008100.1
XP_016863589.1
26
2870
XM_017008101.1
XP_016863590.1
26
3031
XM_017008102.1
XP_016863591.1
25
2846
XM_017008103.1
XP_016863592.1
27
3028
XM_017008104.1
XP_016863593.1
27
2969
XM_017008105.1
XP_016863594.1
26
2945
XM_017008106.1
XP_016863595.1
25
2932
XM_017008107.1
XP_016863596.1
26
2870
XM_017008108.1
XP_016863597.1
25
2830
XM_017008109.1
XP_016863598.1
26
2930
XM_017008110.1
XP_016863599.1
27
2969
XM_017008111.1
XP_016863600.1
26
2945
XM_017008112.1
XP_016863601.1
24
2747
GenBank mRNA
AK095596.1
-
27
2991
AK294720.1
-
6
777
AK299767.1
-
27
2957
AK299815.1
-
27
2969
AK307601.1
-
26
2884
BX537758.1
-
27
2965
X56958.2
-
26
2848
Z26634.2
-
26
2833
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of heart rate
atrial septum development
cellular calcium ion homeostasis
ER to Golgi vesicle-mediated transport
cytoskeleton organization
positive regulation of gene expression
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
regulation of cardiac muscle contraction by calcium ion signaling
paranodal junction assembly
regulation of protein stability
T-tubule organization
protein localization to organelle
protein localization to cell surface
cellular protein localization
protein localization to M-band
protein localization to T-tubule
positive regulation of potassium ion transport
protein stabilization
regulation of release of sequestered calcium ion into cytosol
response to methylmercury
regulation of calcium ion transport
positive regulation of calcium ion transport
regulation of cardiac muscle contraction
regulation of ventricular cardiac muscle cell membrane repolarization
sarcoplasmic reticulum calcium ion transport
protein localization to endoplasmic reticulum
protein localization to plasma membrane
protein targeting to plasma membrane
regulation of cardiac muscle cell contraction
ventricular cardiac muscle cell action potential
atrial cardiac muscle cell action potential
SA node cell action potential
membrane depolarization during SA node cell action potential
atrial cardiac muscle cell to AV node cell communication
SA node cell to atrial cardiac muscle cell communication
regulation of heart rate by cardiac conduction
regulation of SA node cell action potential
regulation of atrial cardiac muscle cell action potential
positive regulation of potassium ion transmembrane transporter activity
regulation of calcium ion transmembrane transporter activity
positive regulation of calcium ion transmembrane transporter activity
positive regulation of cation channel activity
structural constituent of cytoskeleton
protein binding
cytoskeletal adaptor activity
enzyme binding
protein kinase binding
spectrin binding
protein binding, bridging
ion channel binding
ATPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline