Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANK2 SIRNA Assays ›
Gene Symbol:
ANK2
Gene Name:
ankyrin 2, neuronal
Gene Aliases:
ANK-2, LQT4, brank-2
Chromosome Location:
Chr.4: 112706083 - 113383740 on Build GRCh38
UniGene ID:
Hs.620557
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
106391
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127493.1
NP_001120965.1
40
4662
NM_001148.4
NP_001139.3
39
10823
NM_020977.3
NP_066187.2
38
4568
XM_005262945.2
XP_005263002.1
38
10822
XM_017008067.1
XP_016863556.1
41
11780
XM_017008068.1
XP_016863557.1
42
12021
XM_017008069.1
XP_016863558.1
40
11755
XM_017008070.1
XP_016863559.1
41
11966
XM_017008071.1
XP_016863560.1
42
10949
XM_017008072.1
XP_016863561.1
41
10925
XM_017008073.1
XP_016863562.1
39
10921
XM_017008074.1
XP_016863563.1
41
10895
XM_017008075.1
XP_016863564.1
40
10859
XM_017008076.1
XP_016863565.1
39
10921
XM_017008077.1
XP_016863566.1
38
10736
XM_017008078.1
XP_016863567.1
38
4666
XM_017008079.1
XP_016863568.1
36
4396
XM_017008080.1
XP_016863569.1
38
4517
XM_017008081.1
XP_016863570.1
37
4567
XM_017008082.1
XP_016863571.1
39
4744
XM_017008083.1
XP_016863572.1
39
4807
XM_017008084.1
XP_016863573.1
39
4702
XM_017008085.1
XP_016863574.1
39
4663
XM_017008086.1
XP_016863575.1
38
4564
XM_017008087.1
XP_016863576.1
40
4843
XM_017008088.1
XP_016863577.1
41
4637
XM_017008089.1
XP_016863578.1
38
4810
XM_017008090.1
XP_016863579.1
39
4807
XM_017008091.1
XP_016863580.1
37
4504
XM_017008092.1
XP_016863581.1
39
4604
XM_017008093.1
XP_016863582.1
40
4601
XM_017008094.1
XP_016863583.1
39
4663
XM_017008095.1
XP_016863584.1
37
4481
XM_017008096.1
XP_016863585.1
41
4637
XM_017008097.1
XP_016863586.1
38
4442
XM_017008098.1
XP_016863587.1
39
4604
XM_017008099.1
XP_016863588.1
40
4601
XM_017008100.1
XP_016863589.1
37
4406
XM_017008101.1
XP_016863590.1
37
4567
XM_017008102.1
XP_016863591.1
36
4382
XM_017008103.1
XP_016863592.1
39
4663
XM_017008104.1
XP_016863593.1
38
4505
XM_017008105.1
XP_016863594.1
37
4481
XM_017008106.1
XP_016863595.1
36
4468
XM_017008107.1
XP_016863596.1
38
4442
XM_017008108.1
XP_016863597.1
36
4366
XM_017008109.1
XP_016863598.1
36
4433
XM_017008110.1
XP_016863599.1
38
4505
XM_017008111.1
XP_016863600.1
37
4481
XM_017008112.1
XP_016863601.1
35
4283
GenBank mRNA
AK294720.1
-
17
2313
AK299893.1
-
11
1410
BC125235.1
-
6
453
BC125236.1
-
6
453
BX537758.1
-
40
4636
BX538132.1
-
11
1472
BX538239.1
-
3
2728
X56958.2
-
38
4483
Z26634.2
-
39
10723
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of heart rate
atrial septum development
cellular calcium ion homeostasis
ER to Golgi vesicle-mediated transport
cytoskeleton organization
positive regulation of gene expression
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
regulation of cardiac muscle contraction by calcium ion signaling
paranodal junction assembly
regulation of protein stability
T-tubule organization
protein localization to organelle
protein localization to cell surface
cellular protein localization
protein localization to M-band
protein localization to T-tubule
positive regulation of potassium ion transport
protein stabilization
regulation of release of sequestered calcium ion into cytosol
response to methylmercury
regulation of calcium ion transport
positive regulation of calcium ion transport
regulation of cardiac muscle contraction
regulation of ventricular cardiac muscle cell membrane repolarization
sarcoplasmic reticulum calcium ion transport
protein localization to endoplasmic reticulum
protein localization to plasma membrane
protein targeting to plasma membrane
regulation of cardiac muscle cell contraction
ventricular cardiac muscle cell action potential
atrial cardiac muscle cell action potential
SA node cell action potential
membrane depolarization during SA node cell action potential
atrial cardiac muscle cell to AV node cell communication
SA node cell to atrial cardiac muscle cell communication
regulation of heart rate by cardiac conduction
regulation of SA node cell action potential
regulation of atrial cardiac muscle cell action potential
positive regulation of potassium ion transmembrane transporter activity
regulation of calcium ion transmembrane transporter activity
positive regulation of calcium ion transmembrane transporter activity
positive regulation of cation channel activity
structural constituent of cytoskeleton
protein binding
cytoskeletal adaptor activity
enzyme binding
protein kinase binding
spectrin binding
protein binding, bridging
ion channel binding
ATPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-ffltp:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline