Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK1G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022048.3 | NP_071331.2 | 2 | 622 | ||
GenBank mRNA | AB042562.1 | - | 1 | 270 | ||
AB042563.1 | - | 1 | 254 | |||
AF223357.2 | - | 1 | 340 | |||
AK225960.1 | - | 2 | 552 | |||
AL832551.1 | - | 2 | 400 | |||
BC017236.2 | - | 1 | 71 | |||
BC130613.1 | - | 2 | 395 | 40146644 | ||
BC136504.1 | - | 2 | 395 | 9020493 | ||
CF593592.1 | - | 2 | 580 |