Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206.2 | NP_001197.1 | 2 | 3256 | ||
GenBank mRNA | D31716.1 | - | 2 | 3224 |