Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303.3 | NP_001294.2 | 5 | 845 | ||
GenBank mRNA | AK294213.1 | - | 4 | 321 | ||
AK295925.1 | - | 4 | 518 | |||
AK312718.1 | - | 5 | 740 | |||
BC000060.2 | - | 5 | 718 | 3508180 | ||
BC006394.2 | - | 5 | 718 | 4136163 | ||
BT006985.1 | - | 5 | 641 | |||
DA200545.1 | - | 3 | 192 | |||
U09466.1 | - | 5 | 681 |