Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303.3 | NP_001294.2 | 7 | 2814 | ||
GenBank mRNA | BC000060.2 | - | 7 | 2687 | 3508180 | |
BC006394.2 | - | 7 | 2687 | 4136163 | ||
U09466.1 | - | 7 | 2651 |