Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHODH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001361.4 | NP_001352.2 | 9 | 1466 | ||
XM_005255827.3 | XP_005255884.1 | 9 | 1771 | |||
XM_005255829.3 | XP_005255886.1 | 8 | 1408 | |||
XM_017022990.1 | XP_016878479.1 | 9 | 1546 | |||
GenBank mRNA | AK097300.1 | - | 4 | 2110 | ||
BC065245.1 | - | 9 | 1446 | 6064723 | ||
M94065.1 | - | 10 | 1447 |