Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127197.1 | NP_001120669.1 | 6, 7 | 927 | ||
NM_001421.3 | NP_001412.1 | 6, 7 | 1009 | |||
XM_005262389.3 | XP_005262446.1 | 6, 7 | 1345 | |||
XM_011531307.2 | XP_011529609.1 | 5, 6 | 755 | |||
XM_011531308.2 | XP_011529610.1 | 4, 5 | 537 | |||
GenBank mRNA | AF000670.1 | - | 5, 6 | 726 | ||
AK314972.1 | - | 6, 7 | 926 | |||
BC017194.2 | - | 6, 7 | 892 | 3138355 | ||
U32645.1 | - | 6, 7 | 1006 |