Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001460.4 | NP_001451.2 | 5 | 660 | ||
GenBank mRNA | AK312877.1 | - | 5 | 566 | ||
AL833218.1 | - | 5 | 549 | |||
BC005894.1 | - | 5 | 566 | 4293031 | ||
BT006979.1 | - | 4 | 502 | |||
Y09267.1 | - | 5 | 562 |