Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115156.1 | NP_001108628.1 | 4 | 586 | ||
NM_001494.3 | NP_001485.2 | 5 | 721 | |||
XM_017016071.1 | XP_016871560.1 | 5 | 416 | |||
GenBank mRNA | AB451295.1 | - | 5 | 430 | ||
AB451429.1 | - | 5 | 430 | |||
AF144713.1 | - | 2 | 190 | |||
AK297177.1 | - | 6 | 688 | |||
AK297554.1 | - | 4 | 471 | |||
AK309360.1 | - | 6 | 637 | |||
AK312485.1 | - | 5 | 606 | |||
BC005145.2 | - | 5 | 514 | 3504736 | ||
BT006868.1 | - | 5 | 430 | |||
BX648038.1 | - | 4 | 267 | |||
CR457073.1 | - | 5 | 430 | |||
D13988.1 | - | 5 | 450 | |||
EU794614.1 | - | 5 | 430 | |||
Y13286.1 | - | 5 | 582 |