Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286838.1 | NP_001273767.1 | 5 | 820 | ||
NM_001510.3 | NP_001501.2 | 6 | 1105 | |||
XM_011531893.2 | XP_011530195.2 | 5 | 7294 | |||
XM_011531894.2 | XP_011530196.1 | 3 | 333 | |||
XM_011531895.2 | XP_011530197.1 | 3 | 250 | |||
XM_017008118.1 | XP_016863607.1 | 5 | 7294 | |||
XM_017008119.1 | XP_016863608.1 | 5 | 831 | |||
XM_017008120.1 | XP_016863609.1 | 5 | 7294 | |||
XM_017008121.1 | XP_016863610.1 | 5 | 7294 | |||
XM_017008122.1 | XP_016863611.1 | 6 | 1255 | |||
XM_017008123.1 | XP_016863612.1 | 4 | 1609 | |||
XM_017008124.1 | XP_016863613.1 | 3 | 829 | |||
XM_017008125.1 | XP_016863614.1 | 3 | 326 | |||
XM_017008127.1 | XP_016863616.1 | 5 | 7294 | |||
GenBank mRNA | AB209318.1 | - | 6 | 1105 | ||
AF009014.1 | - | 6 | 847 | |||
AK308611.1 | - | 6 | 1019 | |||
BC099652.3 | - | 6 | 847 | 40008396 | ||
BC099653.3 | - | 6 | 847 | 40008397 | ||
BC099654.3 | - | 5 | 562 | 40008398 |