Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL13RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001560.2 | NP_001551.1 | 11 | 3673 | ||
XM_011531336.1 | XP_011529638.1 | 11 | 3951 | |||
XM_011531337.1 | XP_011529639.1 | 4 | 3980 | |||
XM_017029507.1 | XP_016884996.1 | 10 | 3690 | |||
GenBank mRNA | AB209849.1 | - | 11 | 3628 | ||
Y09328.1 | - | 11 | 3663 | |||
Y10659.1 | - | 11 | 3673 |