Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001621.4 | NP_001612.1 | 10 | 2206 | ||
GenBank mRNA | AK307495.1 | - | 10 | 2206 | ||
BC069390.1 | - | 10 | 1663 | 7262381 | ||
BC070080.1 | - | 10 | 2124 | 30342582 | ||
D16354.1 | - | 10 | 2175 | |||
L19872.1 | - | 10 | 1968 |