Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005502.2 | NP_001005502.1 | 3 | 252 | ||
NM_001874.4 | NP_001865.1 | 3 | 284 | |||
NM_198320.3 | NP_938079.1 | 3 | 270 | |||
GenBank mRNA | AF368463.1 | - | 3 | 242 | ||
AK313180.1 | - | 3 | 242 | |||
BC022276.1 | - | 3 | 242 | 4768771 | ||
CB959900.1 | - | 269 | ||||
CB989009.1 | - | 3 | 340 | |||
CD109360.1 | - | 3 | 269 | |||
DA205368.1 | - | 3 | 271 | |||
DB006895.1 | - | 3 | 255 | |||
J04970.2 | - | 3 | 284 |