Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001890.2 | NP_001001890.1 | 6 | 5522 | ||
NM_001754.4 | NP_001745.2 | 9 | 4215 | |||
XM_005261068.3 | XP_005261125.1 | 7 | 4478 | |||
XM_005261069.4 | XP_005261126.1 | 8 | 4059 | |||
XM_011529766.2 | XP_011528068.1 | 9 | 4326 | |||
XM_011529767.2 | XP_011528069.1 | 8 | 4212 | |||
XM_011529768.2 | XP_011528070.1 | 7 | 4020 | |||
XM_017028487.1 | XP_016883976.1 | 7 | 4020 | |||
GenBank mRNA | D43968.1 | - | 6 | 5521 | ||
D43969.1 | - | 8 | 4468 | |||
X90976.1 | - | 1 | 2657 |