Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319145.1 | NP_001306074.1 | 6 | 1536 | ||
NM_001766.3 | NP_001757.1 | 7 | 1815 | |||
XM_005245583.1 | XP_005245640.1 | 6 | 1494 | |||
XM_006711621.1 | XP_006711684.1 | 6 | 1376 | |||
XM_011510125.1 | XP_011508427.1 | 7 | 1630 | |||
XM_011510127.1 | XP_011508429.1 | 6 | 1458 | |||
GenBank mRNA | AK225996.1 | - | 5 | 1250 | ||
BC027926.1 | - | 7 | 1815 | 5229534 | ||
J04142.1 | - | 6 | 1480 |