Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK223393.1 | - | 3 | 619 | ||
AK301154.1 | - | 3 | 642 | |||
AK310676.1 | - | 3 | 481 | |||
AK313614.1 | - | 3 | 648 | |||
BC033583.1 | - | 3 | 603 | 5227138 | ||
CR996443.1 | - | 3 | 625 | |||
M14362.1 | - | 3 | 599 | |||
M16336.1 | - | 3 | 613 | |||
M16445.1 | - | 3 | 596 |