Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323565.1 | NP_001310494.1 | 3, 4 | 614 | ||
NM_001323567.1 | NP_001310496.1 | 2, 3 | 150 | |||
NM_001801.2 | NP_001792.2 | 2, 3 | 557 | |||
NR_136618.1 | 2, 3 | 557 | ||||
NR_136619.1 | 2, 3 | 557 | ||||
NR_136621.1 | 2, 3 | 150 | ||||
GenBank mRNA | AK130357.1 | - | 2, 3 | 519 | ||
AK314231.1 | - | 2, 3 | 507 | |||
AL534761.3 | - | 2, 3 | 556 | |||
BC024241.2 | - | 2, 3 | 507 | 4797463 | ||
BG720517.1 | - | 2, 3 | 155 | |||
BG897741.1 | - | 2, 3 | 202 | |||
CR536540.1 | - | 2, 3 | 246 | |||
D85777.1 | - | 2, 3 | 475 | |||
Z23010.1 | - | 2, 3 | 246 | |||
Z31357.1 | - | 2, 3 | 500 |