Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001804.2 | NP_001795.2 | 3 | 1414 | ||
GenBank mRNA | BC096249.1 | - | 2 | 757 | ||
BC096250.1 | - | 3 | 884 | |||
BC096251.1 | - | 4 | 1031 | |||
BC096252.1 | - | 4 | 1077 | 40005955 | ||
BC127122.1 | - | 4 | 1077 | 40005957 | ||
U15212.1 | - | 3 | 1133 | |||
U51095.1 | - | 3 | 1399 |