Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001917.4 | NP_001908.3 | 11 | 1523 | ||
XM_005268692.3 | XP_005268749.1 | 10 | 1457 | |||
XM_011538004.2 | XP_011536306.1 | 12 | 1609 | |||
XM_011538005.2 | XP_011536307.1 | 12 | 1613 | |||
GenBank mRNA | AX741655.1 | - | 11 | 1513 | ||
AX741656.1 | - | 11 | 1513 | |||
AX741658.1 | - | 10 | 1398 | |||
AX741659.1 | - | 8 | 1285 | |||
AX776478.1 | - | 11 | 1513 | |||
AX776479.1 | - | 11 | 1513 | |||
AX776480.1 | - | 13 | 2442 | |||
AX786386.1 | - | 11 | 1513 | |||
BC029057.1 | - | 11 | 1493 | 5184628 | ||
BM932083.1 | - | 1 | 263 | |||
BX538210.1 | - | 11 | 1455 |