Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159969.1 | NP_001153441.1 | 3, 4 | 692 | ||
NM_001981.2 | NP_001972.1 | 15, 16 | 1561 | |||
XM_005270618.2 | XP_005270675.1 | 14, 15 | 1502 | |||
XM_017000615.1 | XP_016856104.1 | 15, 16 | 1703 | |||
XM_017000619.1 | XP_016856108.1 | 16, 17 | 1585 | |||
XM_017000620.1 | XP_016856109.1 | 15, 16 | 1752 | |||
GenBank mRNA | AK294297.1 | - | 4, 5 | 541 | ||
AK313396.1 | - | 15, 16 | 1510 | |||
BX647676.1 | - | 3, 4 | 673 | |||
CD657119.1 | - | 2, 3 | 336 | |||
DQ367924.1 | - | 15, 16 | 1469 | |||
U07707.1 | - | 15, 16 | 1490 | |||
Z29064.1 | - | 15, 16 | 1561 |