Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EZH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321079.1 | NP_001308008.1 | 20 | 2503 | ||
NM_001321081.1 | NP_001308010.1 | 21 | 2567 | |||
NM_001321082.1 | NP_001308011.1 | 20 | 2474 | |||
NM_001991.4 | NP_001982.2 | 21 | 2594 | |||
XM_005257145.2 | XP_005257202.1 | 19 | 2349 | |||
XM_011524517.2 | XP_011522819.1 | 20 | 2442 | |||
XM_017024350.1 | XP_016879839.1 | 20 | 2421 | |||
XM_017024351.1 | XP_016879840.1 | 19 | 2328 | |||
XM_017024352.1 | XP_016879841.1 | 18 | 2179 | |||
GenBank mRNA | AB002386.1 | - | 20 | 2506 | ||
AK295626.1 | - | 20 | 2457 | |||
AL833159.1 | - | 21 | 2514 | |||
BC015882.1 | - | 21 | 2529 | 3908776 | ||
U50315.1 | - | 20 | 2513 |