Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001888.4 | NP_001879.1 | 6 | 800 | ||
GenBank mRNA | AK290852.1 | - | 4 | 519 | ||
AK310424.1 | - | 4 | 473 | |||
BC018061.1 | - | 4 | 518 | 4817634 | ||
BI860684.1 | - | 5 | 473 | |||
BX648477.1 | - | 6 | 716 | |||
L02950.1 | - | 4 | 479 | |||
U85772.1 | - | 4 | 474 |