Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002028.3 | NP_002019.1 | 12 | 2450 | ||
GenBank mRNA | AK093298.1 | - | 10 | 2208 | ||
BC020232.1 | - | 12 | 2300 | 4652202 | ||
BX248269.1 | - | 12 | 2296 |
Gene Symbol: CHURC1-FNTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202558.1 | NP_001189487.1 | 13 | 2451 | ||
NM_001202559.1 | NP_001189488.1 | 14 | 2522 |