Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002126.4 | NP_002117.1 | 4 | 1911 | ||
XM_005257269.2 | XP_005257326.1 | 4 | 1894 | |||
XM_011524705.1 | XP_011523007.1 | 4 | 1368 | |||
XM_017024556.1 | XP_016880045.1 | 4 | 1365 | |||
GenBank mRNA | AK074289.1 | - | 3 | 1375 | ||
AK290043.1 | - | 4 | 1348 | |||
BC036093.1 | - | 4 | 1865 | 5288156 | ||
M95585.1 | - | 4 | 1708 |