Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002147.3 | NP_002138.1 | 2 | 1538 | ||
GenBank mRNA | BC004454.1 | - | 1 | 467 | ||
BC008940.2 | - | 1 | 467 | |||
M92299.1 | - | 2 | 1755 |