Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171993.1 | NP_001165464.1 | 8 | 821 | ||
NM_002150.2 | NP_002141.1 | 6 | 345 | |||
GenBank mRNA | AK057510.1 | - | 8 | 821 | ||
AK290826.1 | - | 6 | 341 | |||
BC024287.1 | - | 6 | 327 | 5087393 | ||
CD675915.1 | - | 6 | 474 | |||
X72389.1 | - | 6 | 330 |