Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289910.1 | NP_001276839.1 | 11 | 1478 | ||
NM_001290114.1 | NP_001277043.1 | 9 | 1453 | |||
NM_002168.3 | NP_002159.2 | 11 | 1711 | |||
GenBank mRNA | AK222916.1 | - | 11 | 1625 | ||
AK299987.1 | - | 9 | 1367 | |||
BC009244.2 | - | 11 | 1613 | 2959540 | ||
BC071828.1 | - | 11 | 1612 | 6090792 | ||
X69433.1 | - | 11 | 1633 |