Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL9R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002186.2 | NP_002177.2 | 3 | 336 | ||
NM_176786.1 | NP_789743.2 | 4 | 461 | |||
XM_011531151.2 | XP_011529453.1 | 4 | 386 | |||
XM_011531152.2 | XP_011529454.1 | 4 | 362 | |||
XM_011531154.2 | XP_011529456.1 | 4 | 408 | |||
XM_011531155.2 | XP_011529457.1 | 4 | 395 | |||
XM_011531156.2 | XP_011529458.1 | 4 | 393 | |||
XM_011531157.2 | XP_011529459.1 | 4 | 390 | |||
XM_017029495.1 | XP_016884984.1 | 4 | 388 | |||
XM_017029496.1 | XP_016884985.1 | 4 | 396 | |||
XM_017029497.1 | XP_016884986.1 | 4 | 432 | |||
XM_017029498.1 | XP_016884987.1 | 5 | 537 | |||
XM_017029499.1 | XP_016884988.1 | 5 | 459 | |||
XM_017029500.1 | XP_016884989.1 | 4 | 399 | |||
XM_017029501.1 | XP_016884990.1 | 4 | 389 | |||
XM_017029503.1 | XP_016884992.1 | 5 | 438 | |||
XM_017029504.1 | XP_016884993.1 | 4 | 411 | |||
XM_017029505.1 | XP_016884994.1 | 4 | 393 | |||
GenBank mRNA | AK303178.1 | - | 4 | 400 | ||
BC051337.1 | - | 4 | 336 | 6304829 | ||
M84747.1 | - | 3 | 336 |