Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002205.4 | NP_002196.4 | 9 | 962 | ||
GenBank mRNA | AK291570.1 | - | 9 | 907 | ||
AK304696.1 | - | 8 | 785 | |||
AK311051.1 | - | 8 | 714 | |||
AK312414.1 | - | 9 | 962 | |||
AY429549.1 | - | 6 | 351 | |||
BC008786.2 | - | 9 | 947 | 3629647 | ||
M13918.2 | - | 9 | 849 | |||
X06256.1 | - | 9 | 917 |