Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002205.4 | NP_002196.4 | 17, 18 | 1851 | ||
GenBank mRNA | AK291570.1 | - | 17, 18 | 1796 | ||
AK312414.1 | - | 17, 18 | 1851 | |||
BC008786.2 | - | 17, 18 | 1836 | 3629647 | ||
M13918.2 | - | 17, 18 | 1738 | |||
X06256.1 | - | 17, 18 | 1806 |