Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174097.2 | NP_001167568.1 | 7 | 1150 | ||
NM_001315537.1 | NP_001302466.1 | 7 | 928 | |||
NM_002300.7 | NP_002291.1 | 7 | 928 | |||
XM_006719074.2 | XP_006719137.1 | 7 | 1044 | |||
GenBank mRNA | AB209231.1 | - | 1 | 2509 | ||
AK129881.1 | - | 7 | 879 | |||
AK312328.1 | - | 7 | 912 | |||
AY189689.1 | - | 7 | 904 | |||
BC002362.1 | - | 7 | 895 | 2961445 | ||
BC015122.1 | - | 7 | 919 | 3685985 | ||
BC022835.1 | - | 2 | 156 | |||
BC071860.1 | - | 7 | 860 | 4902461 | ||
BT019764.1 | - | 6 | 817 | |||
BT019765.1 | - | 6 | 817 | |||
EU794633.1 | - | 7 | 871 | |||
Y00711.1 | - | 7 | 871 |