Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002396.4 | NP_002387.1 | 14, 15 | 1755 | ||
GenBank mRNA | AJ294818.1 | - | 3, 4 | 236 | ||
AK313391.1 | - | 14, 15 | 1651 | |||
M55905.1 | - | 14, 15 | 1585 |