Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024876.3 | NP_079152.3 | 6 | 775 | ||
GenBank mRNA | AK022291.1 | - | 6 | 775 | ||
BC013114.1 | - | 6 | 585 | 4342891 | ||
CA397752.1 | - | 5 | 421 |