Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289155.1 | NP_001276084.1 | 4, 5 | 560 | ||
NM_001289156.1 | NP_001276085.1 | 3, 4 | 432 | |||
NM_002435.2 | NP_002426.1 | 4, 5 | 560 | |||
XM_011521592.1 | XP_011519894.1 | 4, 5 | 636 | |||
XM_011521593.2 | XP_011519895.1 | 4, 5 | 618 | |||
XM_017022207.1 | XP_016877696.1 | 4, 5 | 640 | |||
XM_017022208.1 | XP_016877697.1 | 4, 5 | 618 | |||
XM_017022209.1 | XP_016877698.1 | 3, 4 | 407 | |||
GenBank mRNA | AF504648.1 | - | 4, 5 | 525 | ||
AK225149.1 | - | 4, 5 | 523 | |||
AK292374.1 | - | 4, 5 | 513 | |||
AK294030.1 | - | 4, 5 | 529 | |||
AK301370.1 | - | 3, 4 | 401 | |||
BC046357.1 | - | 4, 5 | 515 | 5745956 | ||
BX648475.1 | - | 4, 5 | 537 | |||
X76057.1 | - | 4, 5 | 502 |