Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278610.1 | NP_001265539.1 | 5, 6 | 864 | ||
NM_002466.3 | NP_002457.1 | 6, 7 | 936 | |||
GenBank mRNA | AK223482.1 | - | 6, 7 | 671 | ||
AK303249.1 | - | 5, 6 | 769 | |||
AK314791.1 | - | 6, 7 | 841 | |||
BC007585.1 | - | 6, 7 | 861 | 3162656 | ||
BC053555.1 | - | 6, 7 | 830 | 6140320 | ||
X13293.1 | - | 6, 7 | 798 |