Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172623.1 | NP_001166094.1 | 9 | 1680 | ||
NM_001172624.1 | NP_001166095.1 | 9 | 1680 | |||
NM_002499.3 | NP_002490.2 | 9 | 1680 | |||
XM_005254408.1 | XP_005254465.1 | 10 | 1990 | |||
XM_011521628.2 | XP_011519930.1 | 9 | 1745 | |||
XM_011521629.2 | XP_011519931.1 | 9 | 1739 | |||
XM_011521630.2 | XP_011519932.1 | 9 | 1739 | |||
XM_011521632.2 | XP_011519934.1 | 9 | 1739 | |||
XM_011521633.2 | XP_011519935.1 | 9 | 1739 | |||
XM_017022231.1 | XP_016877720.1 | 9 | 1679 | |||
XM_017022232.1 | XP_016877721.1 | 10 | 1990 | |||
XM_017022233.1 | XP_016877722.1 | 9 | 1739 | |||
XM_017022234.1 | XP_016877723.1 | 10 | 1782 | |||
XM_017022235.1 | XP_016877724.1 | 9 | 1739 | |||
XM_017022236.1 | XP_016877725.1 | 9 | 1679 | |||
XM_017022237.1 | XP_016877726.1 | 9 | 1739 | |||
XM_017022238.1 | XP_016877727.1 | 9 | 1679 | |||
GenBank mRNA | AB209412.1 | - | 5 | 576 | ||
BC117161.1 | - | 9 | 1554 | 40125712 | ||
BC143270.1 | - | 9 | 1555 | 9051772 | ||
BC143271.1 | - | 9 | 1554 | 9051773 | ||
U61262.1 | - | 9 | 1624 | |||
U72391.1 | - | 9 | 1614 |