Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEUROD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002500.4 | NP_002491.2 | 2 | 1397 | ||
GenBank mRNA | AB593068.1 | - | 2 | 1249 | ||
AB593069.1 | - | 2 | 1251 | |||
AB593070.1 | - | 2 | 1249 | |||
AB593071.1 | - | 2 | 1249 | |||
BC009046.1 | - | 2 | 1218 | 3873419 | ||
D82347.1 | - | 2 | 1259 |