Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010328.1 | XP_016865817.1 | 1 | 324 |
Gene Symbol: PBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002586.4 | NP_002577.2 | 9 | 3022 | ||
GenBank mRNA | X59842.1 | - | 9 | 3044 |