Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323282.1 | NP_001310211.1 | 4, 5 | 850 | ||
NM_002895.4 | NP_002886.2 | 4, 5 | 850 | |||
NM_183404.3 | NP_899662.1 | 4, 5 | 850 | |||
XM_011528955.2 | XP_011527257.1 | 4, 5 | 626 | |||
XM_011528958.2 | XP_011527260.1 | 4, 5 | 626 | |||
XM_017027992.1 | XP_016883481.1 | 5, 6 | 806 | |||
GenBank mRNA | AK290380.1 | - | 4, 5 | 626 | ||
AK308096.1 | - | 4, 5 | 611 | |||
AK310870.1 | - | 4, 5 | 635 | |||
BC032247.1 | - | 4, 5 | 633 | 4521266 | ||
L14812.1 | - | 4, 5 | 631 | |||
M74547.1 | - | 2, 3 | 164 |