Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185009.2 | NP_001171938.1 | 9 | 2076 | ||
NM_001185010.2 | NP_001171939.1 | 7 | 1394 | |||
NM_002938.4 | NP_002929.1 | 8 | 1554 | |||
GenBank mRNA | AB000468.1 | - | 8 | 1502 | ||
AK128038.1 | - | 8 | 2716 | |||
AK309509.1 | - | 7 | 1322 | |||
AK311314.1 | - | 3 | 882 | |||
BC031935.1 | - | 9 | 2077 | 4824114 | ||
U95140.1 | - | 8 | 1476 |