Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040876.1 | NP_001035809.1 | 7 | 1039 | ||
NM_002940.2 | NP_002931.2 | 7 | 1042 | |||
GenBank mRNA | AB062293.1 | - | 7 | 716 | ||
AK315198.1 | - | 7 | 735 | |||
BC016283.1 | - | 7 | 704 | 3909151 | ||
BT009779.1 | - | 6 | 599 | |||
X76388.1 | - | 7 | 716 |