Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040876.1 | NP_001035809.1 | 18 | 2943 | ||
NM_002940.2 | NP_002931.2 | 18 | 2946 | |||
GenBank mRNA | AB062293.1 | - | 18 | 2620 | ||
BC016283.1 | - | 18 | 2608 | 3909151 | ||
BC016988.1 | - | 10 | 1742 | |||
X74987.1 | - | 12 | 1914 | |||
X76388.1 | - | 18 | 2621 |