Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206974.1 | NP_001193903.1 | 5 | 616 | ||
NM_001930.3 | NP_001921.1 | 5 | 765 | |||
NM_013406.2 | NP_037538.1 | 5 | 765 | |||
NR_038192.1 | 5 | 765 | ||||
XM_011527770.1 | XP_011526072.1 | 5 | 765 | |||
XM_011527771.2 | XP_011526073.1 | 5 | 776 | |||
GenBank mRNA | AK291553.1 | - | 5 | 746 | ||
AK304780.1 | - | 5 | 711 | |||
AL520040.3 | - | 5 | 541 | |||
AY336090.1 | - | 5 | 654 | |||
BC000333.2 | - | 5 | 712 | 2822554 | ||
BC014016.2 | - | 5 | 727 | 4129051 | ||
BU844677.1 | - | 5 | 744 | |||
L39068.1 | - | 5 | 681 | |||
U26266.1 | - | 5 | 646 | |||
U32178.1 | - | 5 | 646 | |||
U40579.1 | - | 5 | 727 | |||
U79262.1 | - | 5 | 739 |